Gene Transcripts

Gene Symbol
Znhit1
Entrez Gene ID
100360077
Gene Name
zinc finger, HIT-type containing 1
Gene Aliases
Chromosome Location
Chr. 12: 22726643 - 22732917 on Build Rnor_6.0
UniGene ID
Rn.2168
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271350.1 NP_001258279.1 2 328
XM_006249091.2 XP_006249153.1 2 203

View Full Product Details