Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001197216.2 | NP_001184145.1 | 8 | 1257 | |
NM_001671.4 | NP_001662.1 | 9 | 1374 | ||
XM_011523861.2 | XP_011522163.1 | 9 | 1281 | ||
GenBank mRNA | AB070933.1 | - | 1153 | ||
AK308375.1 | - | 9 | 1178 | ||
BC032130.2 | - | 9 | 1051 | 5087444 | |
BM818592.1 | - | 1 | 174 | ||
M10058.1 | - | 9 | 1145 |