Gene Transcripts

Gene Symbol
Atxn7
Entrez Gene ID
361015
Gene Name
ataxin 7
Gene Aliases
RGD1562692
Chromosome Location
Chr. 15: 12421533 - 12569650 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006251728.2 XP_006251790.1 9 1456
XM_008770600.1 XP_008768822.1 9 1456

View Full Product Details