Gene Transcripts

Gene Symbol
Alg11
Entrez Gene ID
361174
Gene Name
ALG11, alpha-1,2-mannosyltransferase
Gene Aliases
RGD1564725
Chromosome Location
Chr. 16: 74854006 - 74866252 on Build Rnor_6.0
UniGene ID
Rn.37199
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108401.1 NP_001101871.1 3 610
XM_006253354.2 XP_006253416.1 3 882
XM_006253355.2 XP_006253417.1 3 584
XM_006253356.2 XP_006253418.1 3 1207

View Full Product Details