Gene Transcripts

Gene Symbol
Dok4
Entrez Gene ID
361364
Gene Name
docking protein 4
Gene Aliases
Chromosome Location
Chr. 19: 10563390 - 10575109 on Build Rnor_6.0
UniGene ID
Rn.219052
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108438.2 NP_001101908.1 4 571
XM_008772314.1 XP_008770536.1 3 512
XM_008772315.1 XP_008770537.1 4 604
XM_008772316.1 XP_008770538.1 4 601
XM_008772317.1 XP_008770539.1 3 511
GenBank mRNA BC158749.1 - 4 571

View Full Product Details