Gene Transcripts

Gene Symbol
Edc4
Entrez Gene ID
361399
Gene Name
enhancer of mRNA decapping 4
Gene Aliases
LRRG00115, Nrn1l, RGD1309136
Chromosome Location
Chr. 19: 37852659 - 37864651 on Build Rnor_6.0
UniGene ID
Rn.145173
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033068.3 NP_001028240.3 5, 6 825
GenBank mRNA BC103628.1 - 5, 6 825

View Full Product Details