Gene Transcripts

Gene Symbol
Fam92b
Entrez Gene ID
361423
Gene Name
family with sequence similarity 92, member B
Gene Aliases
RGD1560673
Chromosome Location
Chr. 19: 53740743 - 53754734 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772654.1 XP_008770876.1 4 481

View Full Product Details