Gene Transcripts

Gene Symbol
Ppp6r1
Entrez Gene ID
361502
Gene Name
protein phosphatase 6, regulatory subunit 1
Gene Aliases
RGD1311409, Saps1
Chromosome Location
Chr. 1: 72756774 - 72784492 on Build Rnor_6.0
UniGene ID
Rn.138371
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135849.1 NP_001129321.1 10 1717
XM_006228275.2 XP_006228337.1 10 1976
XM_006228276.2 XP_006228338.1 10 1975

View Full Product Details