Gene Transcripts

Gene Symbol
Vstm2b
Entrez Gene ID
361560
Gene Name
V-set and transmembrane domain containing 2B
Gene Aliases
RGD1306235
Chromosome Location
Chr. 1: 97632473 - 97661541 on Build Rnor_6.0
UniGene ID
Rn.65503
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108479.1 NP_001101949.1 5 1201
XM_008759395.1 XP_008757617.1 3 596

View Full Product Details