Gene Transcripts

Gene Symbol
Prr12
Entrez Gene ID
361569
Gene Name
proline rich 12
Gene Aliases
RGD1309896
Chromosome Location
Chr. 1: 101022374 - 101046205 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759481.1 XP_008757703.1 14 6183

View Full Product Details