Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001108507.2 | NP_001101977.1 | 1, 2 | 148 | |
XM_006230466.2 | XP_006230528.1 | 1, 2 | 2517 | ||
XM_006230469.2 | XP_006230531.1 | 1, 2 | 2516 | ||
GenBank mRNA | BC162015.1 | - | 1, 2 | 148 |