Gene Transcripts

Gene Symbol
Ifitm3
Entrez Gene ID
361673
Gene Name
interferon induced transmembrane protein 3
Gene Aliases
DSPA2b, rat8
Chromosome Location
Chr. 1: 213810712 - 213811821 on Build Rnor_6.0
UniGene ID
Rn.145191
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136124.1 NP_001129596.1 1 189
GenBank mRNA AF164039.1 - 1 166
BC168774.1 - 1 189 6921753

View Full Product Details