Gene Transcripts

Gene Symbol
Tnfrsf26
Entrez Gene ID
361685
Gene Name
tumor necrosis factor receptor superfamily, member 26
Gene Aliases
Chromosome Location
Chr. 1: 216808642 - 216829366 on Build Rnor_6.0
UniGene ID
Rn.162508
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108511.1 NP_001101981.1 3 167
XM_006230827.2 XP_006230889.1 3 952

View Full Product Details