Gene Transcripts

Gene Symbol
Syvn1
Entrez Gene ID
361712
Gene Name
synovial apoptosis inhibitor 1, synoviolin
Gene Aliases
RGD1310488
Chromosome Location
Chr. 1: 221409265 - 221416225 on Build Rnor_6.0
UniGene ID
Rn.162486
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100739.1 NP_001094209.1 5, 6 528
XM_006230852.2 XP_006230914.1 5, 6 970
GenBank mRNA BC166727.1 - 5, 6 528

View Full Product Details