Gene Transcripts

Gene Symbol
Naa40
Entrez Gene ID
361718
Gene Name
N(alpha)-acetyltransferase 40, NatD catalytic subunit
Gene Aliases
Nat11, RGD1565838
Chromosome Location
Chr. 1: 222478021 - 222495382 on Build Rnor_6.0
UniGene ID
Rn.79724
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108518.1 NP_001101988.1 4, 5 494

View Full Product Details