Gene Transcripts

Gene Symbol
Mccc2
Entrez Gene ID
361884
Gene Name
methylcrotonoyl-CoA carboxylase 2 (beta)
Gene Aliases
Chromosome Location
Chr. 2: 30175015 - 30246196 on Build Rnor_6.0
UniGene ID
Rn.33635
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012177.1 NP_001012177.1 10 1086
XM_008760678.1 XP_008758900.1 9 1158
GenBank mRNA BC083581.1 - 10 1086

View Full Product Details