Gene Transcripts

Gene Symbol
Lmo4
Entrez Gene ID
362051
Gene Name
LIM domain only 4
Gene Aliases
Chromosome Location
Chr. 2: 250218635 - 250235435 on Build Rnor_6.0
UniGene ID
Rn.2517
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009708.1 NP_001009708.1 577
XM_006233438.2 XP_006233500.1 343
GenBank mRNA BC087700.1 - 577
FQ233103.1 - 420

View Full Product Details