Gene Transcripts

Gene Symbol
Epb42
Entrez Gene ID
362202
Gene Name
erythrocyte membrane protein band 4.2
Gene Aliases
Epb4.2
Chromosome Location
Chr. 3: 112967095 - 112985318 on Build Rnor_6.0
UniGene ID
Rn.23855
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108590.2 NP_001102060.1 3 331
GenBank mRNA BC168935.1 - 3 331

View Full Product Details