Gene Transcripts

Gene Symbol
Casc4
Entrez Gene ID
362204
Gene Name
cancer susceptibility candidate 4
Gene Aliases
RGD1305985
Chromosome Location
Chr. 3: 113818649 - 113887550 on Build Rnor_6.0
UniGene ID
Rn.55346
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271283.1 NP_001258212.1 1, 2 569

View Full Product Details