Gene Transcripts

Gene Symbol
Csrp2bp
Entrez Gene ID
362224
Gene Name
CSRP2 binding protein
Gene Aliases
Chromosome Location
Chr. 3: 138508899 - 138549437 on Build Rnor_6.0
UniGene ID
Rn.36418
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191855.1 NP_001178784.1 6 1527

View Full Product Details