Gene Transcripts

Gene Symbol
Tamm41
Entrez Gene ID
362419
Gene Name
TAM41, mitochondrial translocator assembly and maintenance protein, homolog (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 4: 146922085 - 146954159 on Build Rnor_6.0
UniGene ID
Rn.210698
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108642.1 NP_001102112.1 5 769
GenBank mRNA FQ213922.1 - 5 739

View Full Product Details