Gene Transcripts

Gene Symbol
Exd2
Entrez Gene ID
362759
Gene Name
exonuclease 3'-5' domain containing 2
Gene Aliases
Exdl2, RGD1311087
Chromosome Location
Chr. 6: 104017620 - 104051442 on Build Rnor_6.0
UniGene ID
Rn.102
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108715.1 NP_001102185.1 2, 3 559
XM_006240286.2 XP_006240348.1 2, 3 907
XM_006240287.2 XP_006240349.1 2, 3 742
GenBank mRNA BC166432.1 - 2, 3 723

View Full Product Details