Gene Transcripts

Gene Symbol
Cmc1
Entrez Gene ID
363162
Gene Name
COX assembly mitochondrial protein 1
Gene Aliases
RGD1305283
Chromosome Location
Chr. 8: 126437345 - 126495347 on Build Rnor_6.0
UniGene ID
Rn.60294
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199225.1 NP_001186154.1 3, 4 356
XM_006244034.2 XP_006244096.1 4, 5 422

View Full Product Details