Gene Transcripts

Gene Symbol
Vgll1
Entrez Gene ID
363508
Gene Name
vestigial-like family member 1
Gene Aliases
RGD1564038
Chromosome Location
Chr. X: 159556296 - 159576639 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773668.1 XP_008771890.1 3 346

View Full Product Details