Gene Transcripts

Gene Symbol
RERE
Entrez Gene ID
473
Gene Name
arginine-glutamic acid dipeptide repeats
Gene Aliases
ARG, ARP, ATN1L, DNB1, NEDBEH
Chromosome Location
Chr. 1: 8352404 - 8826226 on Build GRCh38
UniGene ID
Hs.463041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042681.1 NP_001036146.1 19 4069
NM_001042682.1 NP_001036147.1 9 2368
NM_012102.3 NP_036234.3 20 4254
XM_005263464.2 XP_005263521.1 19 4008
XM_005263466.1 XP_005263523.1 13 2891
XM_011541510.1 XP_011539812.1 18 3994
XM_017001358.1 XP_016856847.1 19 3759
XM_017001359.1 XP_016856848.1 19 4264
GenBank mRNA AB007927.2 - 18 3404
AB036737.1 - 20 4080
AF016005.1 - 9 2346
AF118275.1 - 5 1782

View Full Product Details