Gene Transcripts

Gene Symbol
Phlda3
Entrez Gene ID
363989
Gene Name
pleckstrin homology-like domain, family A, member 3
Gene Aliases
Chromosome Location
Chr. 13: 52588917 - 52592002 on Build Rnor_6.0
UniGene ID
Rn.17905
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012206.2 NP_001012206.1 2 924
GenBank mRNA BC087038.1 - 2 910

View Full Product Details