Gene Transcripts

Gene Symbol
USP13
Entrez Gene ID
8975
Gene Name
ubiquitin specific peptidase 13 (isopeptidase T-3)
Gene Aliases
ISOT3, IsoT-3
Chromosome Location
Chr. 3: 179653145 - 179789401 on Build GRCh38
UniGene ID
Hs.175322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003940.2 NP_003931.2 13 1635
XM_011513269.1 XP_011511571.1 13 1582
XM_017007425.1 XP_016862914.1 13 1479
XM_017007426.1 XP_016862915.1 12 1446
XM_017007427.1 XP_016862916.1 10 1166
GenBank mRNA AK290338.1 - 13 1582
AK302404.1 - 13 1444
BC016146.1 - 13 1588 3910925
HM138083.1 - 13 1554
U75362.1 - 13 1648

View Full Product Details