Gene Transcripts

Gene Symbol
TAF1A
Entrez Gene ID
9015
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit A
Gene Aliases
MGC:17061, RAFI48, SL1, TAFI48
Chromosome Location
Chr. 1: 222557902 - 222590096 on Build GRCh38
UniGene ID
Hs.153088
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001201536.1 NP_001188465.1 6 826
NM_005681.3 NP_005672.1 6 826
NM_139352.2 NP_647603.1 5 656
XM_005273343.3 XP_005273400.1 6 989
XM_006711612.1 XP_006711675.1 5 875
XM_006711613.3 XP_006711676.1 6 839
XM_017002758.1 XP_016858247.1 5 875
XM_017002759.1 XP_016858248.1 6 795
XM_017002760.1 XP_016858249.1 5 603
GenBank mRNA AK001054.1 - 5 508
AK290970.1 - 6 771
AK299536.1 - 6 826
AK315740.1 - 6 810
BC013808.1 - 6 806 3918866
L39060.1 - 5 641

View Full Product Details