Gene Transcripts

Gene Symbol
Xrra1
Entrez Gene ID
365313
Gene Name
X-ray radiation resistance associated 1
Gene Aliases
Chromosome Location
Chr. 1: 164849816 - 164918473 on Build Rnor_6.0
UniGene ID
Rn.138445
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748930.3 XP_003748978.1 3 375
GenBank mRNA BC158768.1 - 3 217

View Full Product Details