Gene Transcripts

Gene Symbol
Trim72
Entrez Gene ID
365377
Gene Name
tripartite motif-containing 72
Gene Aliases
RGD1562778
Chromosome Location
Chr. 1: 199450042 - 199457902 on Build Rnor_6.0
UniGene ID
Rn.23565
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077675.1 NP_001071143.1 4 656
GenBank mRNA BC127539.1 - 4 656
FQ224850.1 - 4 697

View Full Product Details