Gene Transcripts

Gene Symbol
Vom2r47
Entrez Gene ID
365807
Gene Name
vomeronasal 2 receptor, 47
Gene Aliases
Chromosome Location
Chr. 2: 155351173 - 155386987 on Build Rnor_6.0
UniGene ID
Rn.111884
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099506.1 NP_001092976.1 2 766
XM_006232462.1 XP_006232524.1 3 871

View Full Product Details