Gene Transcripts

Gene Symbol
Clvs1
Entrez Gene ID
366311
Gene Name
clavesin 1
Gene Aliases
Clavesin-1, RGD1564200, Rlbp1l1
Chromosome Location
Chr. 5: 22380368 - 22580380 on Build Rnor_6.0
UniGene ID
Rn.168299
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108969.1 NP_001102439.1 3 617
GenBank mRNA FQ214563.1 - 2 442

View Full Product Details