Gene Transcripts

Gene Symbol
Wash
Entrez Gene ID
367328
Gene Name
WAS protein family homolog
Gene Aliases
ORF19, Wash1, Wash2
Chromosome Location
Chr. 9: 113918914 - 113936861 on Build Rnor_6.0
UniGene ID
Rn.15475
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127390.1 NP_001120862.1 1267
GenBank mRNA BC088270.1 - 2 309
BC101888.1 - 5 623
BC166761.1 - 1267 7930951

View Full Product Details