Gene Transcripts

Gene Symbol
Ankib1
Entrez Gene ID
368062
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
Chromosome Location
Chr. 4: 27473225 - 27597206 on Build Rnor_6.0
UniGene ID
Rn.2999
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134781.1 NP_001128253.1 15 2264
XM_006236049.2 XP_006236111.1 15 2516
XM_006236050.2 XP_006236112.1 16 3370
XM_006236051.2 XP_006236113.1 15 1966
XM_008762709.1 XP_008760931.1 15 3488
XM_008762710.1 XP_008760932.1 16 2882
XM_008762711.1 XP_008760933.1 15 3488

View Full Product Details