Gene Transcripts

Gene Symbol
Bst2
Entrez Gene ID
378947
Gene Name
bone marrow stromal cell antigen 2
Gene Aliases
DAMP-1, Damp1
Chromosome Location
Chr. 16: 19938781 - 19942353 on Build Rnor_6.0
UniGene ID
Rn.234068
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198134.2 NP_937767.1 1 230
GenBank mRNA AJ538349.1 - 1 231

View Full Product Details