Gene Transcripts

Gene Symbol
Hyal4
Entrez Gene ID
404783
Gene Name
hyaluronoglucosaminidase 4
Gene Aliases
Chromosome Location
Chr. 4: 52147641 - 52166276 on Build Rnor_6.0
UniGene ID
Rn.124452
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100780.1 NP_001094250.1 3 756
XM_008762799.1 XP_008761021.1 2 784
GenBank mRNA AB100601.1 - 1 345

View Full Product Details