Gene Transcripts

Gene Symbol
Olr34
Entrez Gene ID
404894
Gene Name
olfactory receptor 34
Gene Aliases
Chromosome Location
Chr. 1: 150310319 - 150311257 on Build Rnor_6.0
UniGene ID
Rn.141940
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000689.1 NP_001000689.1 1 765

Gene Symbol
Olr30
Entrez Gene ID
293091
Gene Name
olfactory receptor 30
Gene Aliases
Chromosome Location
Chr. 1: 150225373 - 150226302 on Build Rnor_6.0
UniGene ID
Rn.141942
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000120.1 NP_001000120.1 1 765

View Full Product Details