Gene Transcripts

Gene Symbol
LATS1
Entrez Gene ID
9113
Gene Name
large tumor suppressor kinase 1
Gene Aliases
WARTS, wts
Chromosome Location
Chr. 6: 149658153 - 149718256 on Build GRCh38
UniGene ID
Hs.549084
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270519.1 NP_001257448.1 4 2267
NM_004690.3 NP_004681.1 4 2267
NR_073033.1 5 2173
XM_006715603.3 XP_006715666.1 4 2109
XM_011536252.2 XP_011534554.1 4 2109
XM_017011474.1 XP_016866963.1 4 2109
XM_017011475.1 XP_016866964.1 4 2029
XM_017011476.1 XP_016866965.1 5 2177
XM_017011477.1 XP_016866966.1 4 2109
XM_017011478.1 XP_016866967.1 5 2186
XM_017011479.1 XP_016866968.1 4 2109
XM_017011480.1 XP_016866969.1 3 1806
XM_017011481.1 XP_016866970.1 3 1951
XM_017011482.1 XP_016866971.1 4 2109
GenBank mRNA AF104413.1 - 4 2267
AF164041.1 - 3 1751
AK310422.1 - 5 2173
BC002767.1 - 4 2125 3632571

View Full Product Details