Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003957.1 | NP_001003957.1 | 6, 7 | 898 | |
NM_001003958.1 | NP_001003958.1 | 10, 11 | 1580 | ||
XM_008764530.1 | XP_008762752.1 | 9, 10 | 1439 | ||
XM_008764531.1 | XP_008762753.1 | 6, 7 | 1033 |