Gene Transcripts

Gene Symbol
ZBED1
Entrez Gene ID
9189
Gene Name
zinc finger BED-type containing 1
Gene Aliases
ALTE, DREF, TRAMP, hDREF
Chromosome Location
Chr. Y: 2486414 - 2500967 on Build GRCh38
UniGene ID
Hs.131452
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171135.1 NP_001164606.1 2 1582
NM_001171136.1 NP_001164607.1 2 1504
NM_004729.3 NP_004720.1 2 1501
GenBank mRNA AB018328.1 - 2 1501
BC015030.1 - 2 1582 3908412
Y16947.1 - 1 1356

View Full Product Details