Gene Transcripts

Gene Symbol
Vom1r67
Entrez Gene ID
494263
Gene Name
vomeronasal 1 receptor 67
Gene Aliases
V1rc20
Chromosome Location
Chr. 4: 87353254 - 87354165 on Build Rnor_6.0
UniGene ID
Rn.129673
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008912.1 NP_001008912.1 1 501
GenBank mRNA AY510295.1 - 1 501

View Full Product Details