Gene Transcripts

Gene Symbol
Vom1r73
Entrez Gene ID
494271
Gene Name
vomeronasal 1 receptor 73
Gene Aliases
V1rc34
Chromosome Location
Chr. 4: 87758181 - 87759098 on Build Rnor_6.0
UniGene ID
Rn.129667
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008922.1 NP_001008922.1 1 871
GenBank mRNA AY510289.1 - 1 871

View Full Product Details