Gene Transcripts

Gene Symbol
Vom1r98
Entrez Gene ID
494294
Gene Name
vomeronasal 1 receptor 98
Gene Aliases
V1ra8
Chromosome Location
Chr. 4: 121912352 - 121913296 on Build Rnor_6.0
UniGene ID
Rn.129644
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008948.1 NP_001008948.1 1 866
GenBank mRNA AY510365.1 - 1 866

View Full Product Details