Gene Transcripts

Gene Symbol
Vom1r14
Entrez Gene ID
494304
Gene Name
vomeronasal 1 receptor 14
Gene Aliases
V1re7
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.129453
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008961.1 NP_001008961.1 1 490
GenBank mRNA AY510357.1 - 1 490

Gene Symbol
Vom1r12
Entrez Gene ID
494306
Gene Name
vomeronasal 1 receptor 12
Gene Aliases
V1re23
Chromosome Location
Chr. 1: 60406375 - 60407295 on Build Rnor_6.0
UniGene ID
Rn.129454
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008962.1 NP_001008962.1 1 475
GenBank mRNA AY510364.1 - 1 475

Gene Symbol
Vom1r15
Entrez Gene ID
494305
Gene Name
vomeronasal 1 receptor 15
Gene Aliases
V1re22
Chromosome Location
Chr. 1: 60633626 - 60634546 on Build Rnor_6.0
UniGene ID
Rn.129452
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008960.1 NP_001008960.1 1 475
GenBank mRNA AY510361.1 - 1 475

View Full Product Details