Gene Transcripts

Gene Symbol
Myo19
Entrez Gene ID
497974
Gene Name
myosin XIX
Gene Aliases
RGD1559961
Chromosome Location
Chr. 10: 72198475 - 72227892 on Build Rnor_6.0
UniGene ID
Rn.17552
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163736.1 NP_001157208.1 17 1903
XM_008768098.1 XP_008766320.1 17 2035
XM_008768099.1 XP_008766321.1 17 2034

View Full Product Details