Gene Transcripts

Gene Symbol
Med1
Entrez Gene ID
497991
Gene Name
mediator complex subunit 1
Gene Aliases
RGD1559552
Chromosome Location
Chr. 10: 86101560 - 86145274 on Build Rnor_6.0
UniGene ID
Rn.4262
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134361.1 NP_001127833.1 13 1140
XM_006247398.2 XP_006247460.1 13 1290
XM_006247399.2 XP_006247461.1 14 1365
XM_006247401.2 XP_006247463.1 13 1289
XM_008768105.1 XP_008766327.1 13 1920

View Full Product Details