Gene Transcripts

Gene Symbol
DLGAP1
Entrez Gene ID
9229
Gene Name
DLG associated protein 1
Gene Aliases
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1
Chromosome Location
Chr. 18: 3496032 - 4455307 on Build GRCh38
UniGene ID
Hs.594640
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003809.2 NP_001003809.1 5 773
NM_001242761.1 NP_001229690.1 8 2130
NM_001242763.1 NP_001229692.1 6 830
NM_001242764.1 NP_001229693.1 5 746
NM_001242765.1 NP_001229694.1 5 773
NM_001242766.1 NP_001229695.1 6 803
NM_001308390.1 NP_001295319.1 6 828
NM_004746.3 NP_004737.2 8 2130
XM_005258171.2 XP_005258228.1 9 2201
XM_005258172.1 XP_005258229.1 9 2196
XM_005258173.3 XP_005258230.2 7 992
XM_005258174.4 XP_005258231.1 6 1049
XM_006722367.3 XP_006722430.1 7 1160
XM_017026080.1 XP_016881569.1 6 1129
XM_017026081.1 XP_016881570.1 5 793
XM_017026082.1 XP_016881571.1 6 948
XM_017026083.1 XP_016881572.1 5 1018
XM_017026084.1 XP_016881573.1 5 1015
XM_017026085.1 XP_016881574.1 5 1008
GenBank mRNA AB000276.1 - 5 716
AB000277.1 - 5 1670
AK294747.1 - 6 803
AK294777.1 - 6 753
AK299880.1 - 5 746
AK304101.1 - 8 2112
AK316099.1 - 8 2130
BC136453.1 - 5 1662 9020442
BC136454.1 - 5 1662 9020443
U67988.1 - 5 720

View Full Product Details