Gene Transcripts

Gene Symbol
Wdr26
Entrez Gene ID
498301
Gene Name
WD repeat domain 26
Gene Aliases
RGD1565589
Chromosome Location
Chr. 13: 99493120 - 99531959 on Build Rnor_6.0
UniGene ID
Rn.23078
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109081.1 NP_001102551.1 5 529
GenBank mRNA FQ226610.1 - 523
FQ227016.1 - 523

View Full Product Details