Gene Transcripts

Gene Symbol
Cytl1
Entrez Gene ID
498392
Gene Name
cytokine like 1
Gene Aliases
RGD1561846
Chromosome Location
Chr. 14: 77805137 - 77810147 on Build Rnor_6.0
UniGene ID
Rn.8720
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134604.1 NP_001128076.1 2 196
GenBank mRNA BC169053.1 - 2 196

View Full Product Details