Gene Transcripts

Gene Symbol
Defb52
Entrez Gene ID
498660
Gene Name
defensin beta 52
Gene Aliases
Chromosome Location
Chr. 16: 75572014 - 75574546 on Build Rnor_6.0
UniGene ID
Rn.125328
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037524.1 NP_001032613.1 3 137
XM_006253368.2 XP_006253430.1 3 193
GenBank mRNA DQ012091.1 - 3 137

View Full Product Details